5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide

Synonyms: 5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulfonamidePazopanib5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulphonamide

1916-5 1916-25
10188-498EA 274.36 USD
10188-498 10188-496
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide
Formula: C₂₁H₂₃N₇O₂S
MW: 437.53 g/mol
MDL Number: MFCD11616589
CAS Number: 444731-52-6

Order Now

Specification Test Results

CAS 444731-52-6
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR