Cyclopamine-KAAD

Supplier: BioVision

AB142146-50UG
10188-476EA 306.43 USD
10188-476
Cyclopamine-KAAD
Cyclopamine-KAAD
CAS Number: 306387-90-6

Order Now

Specification Test Results

CAS 306387-90-6
Purity ≥80% by HPLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR