GYY 4137

Supplier: BioVision

AB142145-5MG AB142145-25MG
10188-468EA 160.9 USD
10188-468 10188-470
GYY 4137
GYY 4137
Formula: C₁₁H₁₆NO₂PS₂·C₄H₉NO
CAS Number: 106740-09-4

Order Now

Specification Test Results

CAS 106740-09-4
Purity ≥95% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR