TG003
Supplier: BioVision
Synonyms:
1-(3-Ethyl-5-methoxybenzo[d]thiazol-2(3H)-ylidene)propan-2-one
AB141463-5MG
AB141463-25MG
10188-458EA
244.79
USD
10188-458
10188-460
TG003
TG003
Formula:
C₁₃H₁₅NO₂S MW: 249.33 g/mol |
MDL Number:
MFCD00624584 CAS Number: 300801-52-9 |
Specification Test Results
CAS | 300801-52-9 |
Purity | ≥98% by TLC |
Form | Pale yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...