Kenpaullone
Synonyms:
9-Bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one
, 9-Bromo-7,12-dihydrobenzo[2,3]azepino[4,5-b]indol-6(5H)-one
1904-1
10188-456EA
93.39
USD
10188-456
Kenpaullone
Kenpaullone
Formula:
C₁₆H₁₁BrN₂O MW: 327.18 g/mol |
MDL Number:
MFCD02683595 CAS Number: 142273-20-9 |
Specification Test Results
CAS | 142273-20-9 |
Purity | ≥98% |
Form | Tan solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...