SR1001 ≥98.0% (by TLC, NMR)

Supplier: BioVision

Synonyms: N-(5-(N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)sulfamoyl)-4-methylthiazol-2-yl)acetamideN-(5-(N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)sulphamoyl)-4-methylthiazol-2-yl)acetamide

AB141462-5MG AB141462-25MG
10188-448EA 245.16 USD
10188-448 10188-450
SR1001 ≥98.0% (by TLC, NMR)
SR1001

A high affinity synthetic ligand to retinoic acid-related orphan receptors ɑ (RORɑ) and gamma

Formula: C₁₅H₁₃F₆N₃O₄S₂
MW: 477.40 g/mol
MDL Number: MFCD23160036
CAS Number: 1335106-03-0

Order Now

Specification Test Results

Purity ≥98% by TLC and NMR
Form Cream solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR