Dynasore

Synonyms: 3-Hydroxynaphthalene-2-carboxylic acid (3,4-Dihydroxybenzylidene)hydrazideN'-(3,4-Dihydroxybenzylidene)-3-hydroxy-2-naphthohydrazide

1900-25 1900-5
10188-444EA 76.82 USD
10188-444 10188-446
Dynasore
Dynasore
Formula: C₁₈H₁₄N₂O₄
MW: 322.32 g/mol
MDL Number: MFCD00292551
CAS Number: 304448-55-3

Order Now

Specification Test Results

CAS 304448-55-3
Purity ≥98% by NMR
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR