DPQ

Supplier: BioVision

AB141461-1MG AB141461-5MG
10188-440EA 191.47 USD
10188-440 10188-442
DPQ
DPQ
Formula: C₁₈H₂₆N₂O₂
MW: 302.4
MDL Number: MFCD01866274
CAS Number: 129075-73-6

Order Now

Specification Test Results

CAS 129075-73-6
Purity ≥99% by NMR
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR