Alsterpaullone

Synonyms: 7,12-Dihydro-9-nitroindolo[3,2-d][1]benzazepin-6(5H)-one

1897-5 1897-1
10188-432EA 110.91 USD
10188-432 10188-434
Alsterpaullone
Alsterpaullone
Formula: C₁₆H₁₁N₃O₃
MW: 293.3 g/mol
MDL Number: MFCD02683579
CAS Number: 237430-03-4

Order Now

Specification Test Results

CAS 237430-03-4
Purity ≥95% by HPLC
Form Yellow to brown solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR