Tomatidine HCl
1893-25
10191-162EA
93.39
USD
10191-162
Tomatidine HCl
Tomatidine HCl
Formula:
C₂₇H₄₅NO₂·HCl |
CAS Number:
6192-62-7 |
Specification Test Results
CAS | 6192-62-7 |
Purity | ≥90% |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...