GANT61

Synonyms: 2,2'-((2-(Pyridin-4-yl)dihydropyrimidine-1,3(2H,4H)-diyl)bis(methylene))bis(N,N-dimethylaniline)

1892-5
10191-160EA 192.64 USD
10191-160
GANT61
GANT61
Formula: C₂₇H₃₅N₅
MW: 429.6 g/mol
MDL Number: MFCD14635408
CAS Number: 500579-04-4

Order Now

Specification Test Results

CAS 500579-04-4
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR