GDC-0449

Synonyms: 2-Chloro-N-(4-chloro-3-(pyridin-2-yl)phenyl)-4-(methylsulphonyl)benzamideVismodegib2-Chloro-N-(4-chloro-3-(pyridin-2-yl)phenyl)-4-(methylsulfonyl)benzamide

1890-5 1890-25
10191-154EA 607.09 USD
10191-154 10188-422
GDC-0449
GDC-0449
Formula: C₁₉H₁₄Cl₂N₂O₃S
MW: 421.3 g/mol
MDL Number: MFCD12407408
CAS Number: 879085-55-9

Order Now

Specification Test Results

CAS 879085-55-9
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR