Phenethylbiguanide hydrochloride

Supplier: BioVision

Synonyms: 1-Carbamimidamido-N-(2-phenylethyl)methanimidamide Hydrochloride

AB141459-100MG 1889-1000
10188-420EA 332.22 USD
10188-420 10188-418
Phenethylbiguanide hydrochloride
Phenethylbiguanide hydrochloride
Formula: C₁₀H₁₆ClN₅
MW: 241.7205 g/mol
MDL Number: MFCD00035039
CAS Number: 834-28-6

Order Now

Specification Test Results

CAS 834-28-6
Purity ≥97% by TLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR