Pimozide

Supplier: BioVision

AB142135-50MG AB142135-250MG
10188-410EA 191.23 USD
10188-410 10188-412
Pimozide
Pimozide
CAS Number: 2062-78-4

Order Now

Specification Test Results

CAS 2062-78-4
Purity ≥98% by TLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR