Mycophenolic acid ≥98%

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

1882-50 1882-250
10188-390EA 69.94 USD
10188-390 10188-392
Mycophenolic acid ≥98%
Mycophenolic acid

An Immunosuppressive agent that displays antiviral and antitumor effects in vitro and in vivo


  • White to off-white solid


Potently inhibits inosine monophosphate dehydrogenase in the guanosine monophosphate pathway. Also suppresses cytokine-induced nitric oxide production.


Soluble in DMSO (100 mM) or EtOH (100 mM)


Caution: For research use only, not for use in humans.

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR