Glibenclamide

Synonyms: 5-Chloro-N-[4-(3-cyclohexylureidosulphonyl)phenethyl]-2-methoxybenzamideGlyburide5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulphamoyl)phenethyl)-2-methoxybenzamide5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulfamoyl)phenethyl)-2-methoxybenzamide

1878-500 1878-1000
10191-694EA 81.72 USD
10191-694 10191-696
Glibenclamide
Glibenclamide
Formula: C₂₃H₂₈ClN₃O₅S
MW: 494.00 g/mol
MDL Number: MFCD00056625
CAS Number: 10238-21-8

Order Now

Specification Test Results

CAS 10238-21-8
Purity ≥99% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR