Salermide
Synonyms:
N-(3-(((2-Hydroxynaphthalen-1-yl)methylene)amino)phenyl)-2-phenylpropanamide
1873-5
1873-25
10188-368EA
116.75
USD
10188-368
10188-370
Salermide
Salermide
Formula:
C₂₆H₂₂N₂O₂ MW: 394.47 g/mol |
MDL Number:
MFCD12912446 CAS Number: 1105698-15-4 |
Specification Test Results
CAS | 1105698-15-4 |
Purity | ≥98% |
Form | Yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...