PD 184,352 , EZSolution™

Synonyms: 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamideCI-1040PD 1843522-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide

1871-1
10188-364EA 93.39 USD
10188-364
PD 184,352 , EZSolution™
PD 184,352
Formula: C₁₇H₁₄ClF₂IN₂O₂
MW: 478.7 g/mol
MDL Number: MFCD02683961
CAS Number: 212631-79-3

Order Now

Specification Test Results

CAS 212631-79-3
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR