Rocaglamide

Supplier: BioVision

Synonyms: (1R,2R,3S,3aR,8bS)-1,8b-Dihydroxy-6,8-dimethoxy-3a-(4-methoxyphenyl)-N,N-dimethyl-3-phenyl-2,3,3a,8b-tetrahydro-1H-cyclopenta[b]benzofuran-2-carboxamide

AB142130-100UG
10191-684EA 903.99 USD
10191-684
Rocaglamide
Rocaglamide
Formula: C₂₉H₃₁NO₇
MW: 505,56 g/mol
MDL Number: MFCD08702699
CAS Number: 84573-16-0

Order Now

Specification Test Results

CAS 84573-16-0
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR