Puromycin dihydrochloride ≥98% (by HPLC), EZSolution™
Supplier: BioVision
AB146610-100MG
10188-332EA
532.68
USD
10188-332
Puromycin dihydrochloride ≥98% (by HPLC), EZSolution™
Puromycin dihydrochloride
A solution of Puromycin Dihydrochloride in deionized water
- Liquid
Suitable for cell culture applications.
H2O (10 mg/mL, 0.2 µM sterile-filtered)
Caution: For research use only, not for use in humans.
Specification Test Results
CAS | 58-58-2 |
Purity | ≥98% by HPLC |
Form | Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...