DBeQ

Synonyms: N,N'-Dibenzylquinazoline-2,4-diamineN2,N4-Dibenzylquinazoline-2,4-diamine

1858-25 1858-5
10188-326EA 110.91 USD
10188-326 10191-142
DBeQ
DBeQ
Formula: C₂₂H₂₀N₄
MW: 340.43 g/mol
MDL Number: MFCD03691820
CAS Number: 177355-84-9

Order Now

Specification Test Results

CAS 177355-84-9
Purity ≥98% by HPLC
Form Colorless solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR