SMER28

Supplier: BioVision

Synonyms: N-Allyl-6-bromoquinazolin-4-amine

AB142127-25MG AB142127-5MG
10188-318EA 321.66 USD
10188-318 10188-320
SMER28
SMER28
Formula: C₁₁H₁₀BrN₃
MW: 264.126 g/mol
MDL Number: MFCD02166825
CAS Number: 307538-42-7

Order Now

Specification Test Results

CAS 307538-42-7
Purity ≥98% by TLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR