DAPT ≥98% (by TLC)

Synonyms: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Estertert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate(3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester(3,5-Difluorophenylacetyl)-Ala-Phg-OtBu

1855-5
10188-316EA 220.57 USD
10188-316
DAPT ≥98% (by TLC)
DAPT

Cell permeable. A potent inhibitor of γ-secretase (IC₅₀ = 115 nM for total β-amyloid, IC₅₀ = 200 nM for β-amyloid 1-42). It reduces Aβ levels in vivo in plasma and cerebrospinal fluid in young and aged Tg2576 mice. Antagonizes Notch signaling through inhibition of Notch processing. DAPT treatment can influence hematopoietic cell fate decisions and enhances neuronal differentiation in embryonic stem cell-derived embryoid bodies independent of sonic hedgehog (shh) signaling.


  • Appearance: White to off-white solid

Formula: C₂₃H₂₆F₂N₂O₄
MW: 432.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04974585
CAS Number: 208255-80-5

Order Now

Specification Test Results

CAS 208255-80-5
Purity ≥98% by TLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR