Curcumin (from Curcuma longa), high purity

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

1850-10 1850-50
10188-296EA 70.04 USD
10188-296 10188-298
Curcumin (from Curcuma longa), high purity
Curcumin
Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
MDL Number: MFCD00008365
CAS Number: 458-37-7

Order Now

Specification Test Results

CAS 458-37-7
Purity ≥98%
Form Orange-yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR