Bortezomib

Synonyms: ((R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butyl)boronic acid

1846-1 1846-5
10188-286EA 286.04 USD
10188-286 10188-284
Bortezomib
Bortezomib
Formula: C₁₉H₂₅BN₄O₄
MW: 384.24 g/mol
MDL Number: MFCD09056737
CAS Number: 179324-69-7

Order Now

Specification Test Results

CAS 179324-69-7
Purity ≥99% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR