Rosmarinic acid

Supplier: BioVision

Synonyms: (R,E)-3-(3,4-Dihydroxyphenyl)-2-((3-(3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid

AB141450-50 AB141450-250MG
10188-274EA 444.35 USD
10188-274 10188-272
Rosmarinic acid
Rosmarinic acid
Formula: C₁₈H₁₆O₈
MW: 360.318 g/mol
MDL Number: MFCD00017740
CAS Number: 20283-92-5

Order Now

Specification Test Results

CAS 20283-92-5
Purity ≥95% by HPLC
Form Beige solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR