(-)-Epigallocatechin gallate

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate(2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoateEGCG

1841-50
10188-266EA 110.91 USD
10188-266
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate
Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
MDL Number: MFCD00075940
CAS Number: 989-51-5

Order Now

Specification Test Results

CAS 989-51-5
Purity ≥98% by HPLC
Form Off-white to yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR