8-Bromo-cAMP sodium salt
Synonyms:
8-Br-cAMP
, 8-Bromo-cAMP
, 8- Bromoadenosine- 3', 5'-monophosphate
, 8- Bromoadenosine- 3', 5'- cyclic monophosphate
, 8-Br-Camp sodium salt
1837-5
1837-25
10191-140EA
75.87
USD
10191-140
10188-256
8-Bromo-cAMP sodium salt
8-Bromo-cAMP sodium salt
Formula:
C₁₀H₁₀BrN₅NaO₆P MW: 430.08 g/mol |
MDL Number:
MFCD00005844 CAS Number: 76939-46-3 |
Specification Test Results
CAS | 76939-46-3 |
Purity | ≥98% by TLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...