Niclosamide
1826-500
1826-100
10188-228EA
104.17
USD
10188-228
10188-230
Niclosamide
Niclosamide
Formula:
C₁₃H₈Cl₂N₂O₄ |
MDL Number:
MFCD00057597 CAS Number: 50-65-7 |
Specification Test Results
CAS | 50-65-7 |
Purity | ≥98% by HPLC |
Form | Off-white to pale yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...