SH-5

Supplier: BioVision

AB141442-500UG
10188-202EA 543.93 USD
10188-202
SH-5
SH-5
Formula: C₂₉H₅₉O₁₀P
MW: 598.75 g/mol
MDL Number: MFCD08703608
CAS Number: 701976-54-7

Order Now

Specification Test Results

Purity ≥98%
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR