Cp 31398 Dihydrochloride

Synonyms: N'-[2-[2-(4-Methoxyphenyl)ethenyl]-4-quinazolinyl]-N,N-dimethyl-1,3-propanediamine dihydrochloride

1815-25 1815-5
10188-190EA 426.14 USD
10188-190 10188-188
Cp 31398 Dihydrochloride
Cp 31398 Dihydrochloride
Formula: C₂₂H₂₈Cl₂N₄O
MW: 435.393 g/mol
MDL Number: MFCD11519956
CAS Number: 259199-65-0

Order Now

Specification Test Results

CAS 1217195-61-3
Purity ≥98% by TLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR