GANT58

Synonyms: 4,4',4'',4'''-(Thiophene-2,3,4,5-tetrayl)tetrapyridine

1812-5 1812-25
10187-888EA 426.14 USD
10187-888 10187-886
GANT58
GANT58
Formula: C₂₄H₁₆N₄S
MW: 392.4756 g/mol
MDL Number: MFCD01103196
CAS Number: 64048-12-0

Order Now

Specification Test Results

CAS 64048-12-0
Purity ≥98% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR