IKK-ε Kinase inhibitor II

Supplier: BioVision

Synonyms: 5-(5,6-dimethoxy-1H-benzimidazol-1-yl)-3-[[2-(methylsulfonyl)phenyl]methoxy]-2-thiophenecarbonitrileCAY10576IKK-35-(5,6-dimethoxy-1H-benzimidazol-1-yl)-3-[[2-(methylsulphonyl)phenyl]methoxy]-2-thiophenecarbonitrile

AB142109-1MG AB142109-5MG
10187-884EA 995.92 USD
10187-884 10188-182
IKK-ε Kinase inhibitor II
IKK-ε Kinase
Formula: C₂₂H₁₉N₃O₅S₂
MW: 469.5 g/mol
CAS Number: 862812-98-4

Order Now

Specification Test Results

CAS 862812-98-4
Purity ≥97% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR