Diphenylboric acid-2-aminoethyl ester (2-APB)

Synonyms: 2-Aminoethyl diphenylborinate2-((Diphenylboryl)oxy)ethanamine

1798-100
10188-162EA 70.89 USD
10188-162
Diphenylboric acid-2-aminoethyl ester (2-APB)
Diphenylboric acid 2-aminoethyl ester (2-APB)
Formula: C₁₄H₁₆BNO
MW: 225.1 g/mol
MDL Number: MFCD00014823
CAS Number: 524-95-8

Order Now

Specification Test Results

CAS 524-95-8
Purity ≥97%
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR