BAY 61-3606 hydrochloride
Supplier: BioVision
AB142106-5MG
AB142106-1MG
10188-154EA
268.14
USD
10188-154
10188-156
BAY 61-3606 hydrochloride
BAY 61-3606 hydrochloride
Formula:
C₂₀H₁₈N₆O₃·HCl |
CAS Number:
732938-37-8 |
Specification Test Results
CAS | 732983-37-8 |
Purity | ≥98% by HPLC |
Form | Yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...