Fasudil hydrochloride

Synonyms: 5-(1-Homopiperazinyl)sulphonylisoquinoline Hydrochloride5-((1,4-Diazepan-1-yl)sulphonyl)isoquinoline hydrochloride5-((1,4-Diazepan-1-yl)sulfonyl)isoquinoline hydrochloride1-(5-Isoquinolinesulphonyl)homopiperazine Hydrochloride1-(5-Isoquinolinesulfonyl)homopiperazine Hydrochloride5-(1-Homopiperazinyl)sulfonylisoquinoline Hydrochloride

1787-10 1787-50
10187-882EA 119.1 USD
10187-882 10188-134
Fasudil hydrochloride
Fasudil hydrochloride
Formula: C₁₄H₁₇N₃O₂S
MW: 327.83 g/mol
MDL Number: MFCD00943198
CAS Number: 105628-07-7

Order Now

Specification Test Results

CAS 105628-07-7
Purity ≥99% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR