SB203580 , EZSolution™

Synonyms: 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine4-(5-(4-Fluorophenyl)-2-(4-(methylsulfinyl)phenyl)-1H-imidazol-4-yl)pyridine

1786-1
10187-880EA 93.39 USD
10187-880
SB203580 , EZSolution™
SB203580
Formula: C₂₁H₁₆FN₃OS
MW: 377.4 g/mol
MDL Number: MFCD00922198
CAS Number: 152121-47-6

Order Now

Specification Test Results

CAS 152121-47-6
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR