Octyl-β-D-thioglucopyranoside

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol1-S-Octyl-β-D-thioglucopyranosideOctyl-Thioglucoside

1782-50G 1782-1G 1782-100G 1782-5G
10187-868EA 326.9 USD
10187-868 10187-866 10187-872 10187-870
Octyl-β-D-thioglucopyranoside
Octyl β-D-thioglucopyranoside
Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now

Specification Test Results

CAS 85618-21-9
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR