AS-605240

Synonyms: 5-(Quinoxalin-6-ylmethylene)thiazolidine-2,4-dione

1780-1 1780-5
10187-858EA 87.57 USD
10187-858 10187-860
AS-605240
AS-605240
Formula: C₁₂H₇N₃O₂S
MW: 257.2679 g/mol
MDL Number: MFCD11114066
CAS Number: 648450-29-7

Order Now

Specification Test Results

CAS 648450-29-7
Purity ≥98% by HPLC
Form Pink to reddish solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR