N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16)
Synonyms:
Hexadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt
, Palmityl sulphobetaine
, Palmityl sulfobetaine
, 3-(Hexadecyldimethylammonio)propane-1-sulphonate
, Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt
, 3-[Dimethyl(palmityl)ammonio]propane-1-sulfonate
, 3-[Dimethyl(palmityl)ammonio]propane-1-sulphonate
, Palmityldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt
, Palmityldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt
, 3-(Hexadecyldimethylammonio)propane-1-sulfonate
1777-25G
1777-5G
10188-132EA
104.92
USD
10188-132
10187-848
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16)
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16)
Specification Test Results
CAS | 2281-11-0 |
Purity | ≥98% by TLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...