S3I-201

Supplier: BioVision

Synonyms: 2-Hydroxy-4-(2-(tosyloxy)acetamido)benzoic acid

AB141434-5MG AB141434-25MG
10188-124EA 222.17 USD
10188-124 10188-126
S3I-201
S3I-201
Formula: C₁₆H₁₅NO₇S
MW: 365,36 g/mol
MDL Number: MFCD09907564
CAS Number: 501919-59-1

Order Now

Specification Test Results

CAS 501919-59-1
Purity ≥97% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR