17-AAG

Synonyms: 17-(Allylamino)-17-demethoxygeldanamycin(4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

1774-1 1774-5
10188-120EA 163.2 USD
10188-120 10188-122
17-AAG
17-AAG
Formula: C₃₁H₄₃N₃O₈
MW: 585.7 g/mol
MDL Number: MFCD04973892
CAS Number: 75747-14-7

Order Now

Specification Test Results

CAS 75747-14-7
Purity ≥98% by HPLC
Form Purple solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR