D4476

Synonyms: 4-(4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl)benzamide

1770-1 1770-5
10188-106EA 361.93 USD
10188-106 10188-104
D4476
D4476
Formula: C₂₃H₁₈N₄O₃
MW: 398.4140 g/mol
MDL Number: MFCD09037526
CAS Number: 301836-43-1

Order Now

Specification Test Results

CAS 301836-43-1
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR