H-89 dihydrochloride
1768-5
1768-1
10188-098EA
431.97
USD
10188-098
10188-096
H-89 dihydrochloride
H-89 dihydrochloride
Formula:
C₂₀H₂₀BrN₃O₂S·2HCl |
CAS Number:
127243-85-0 |
Specification Test Results
CAS | 127243-85-0 |
Purity | ≥98% by HPLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...