PP2

Synonyms: 1-(tert-Butyl)-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

1767-1 1767-5
10188-092EA 116.75 USD
10188-092 10188-094
PP2
PP2
Formula: C₁₅H₁₆ClN₅
MW: 301.8 g/mol
MDL Number: MFCD01568210
CAS Number: 172889-27-9

Order Now

Specification Test Results

CAS 172889-27-9
Purity ≥98% by HPLC
Form Cream solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR