Okadaic acid sodium salt

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium saltOA sodium salt

1764-025 1764-100
10188-080EA 122.58 USD
10188-080 10188-082
Okadaic acid sodium salt
Okadaic acid sodium salt
Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
MDL Number: MFCD00210208
CAS Number: 209266-80-8

Order Now

Specification Test Results

CAS 209266-80-8
Purity ≥97% by TLC
Form White solid or clear, colorless film

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR