Fludarabine monophosphate

Synonyms: Fludarabine Phosphate2-F-ara-AMP9-(beta-D-Arabinofuranosyl)-2-fluoroadenine 5'-Monophosphate2-Fluoroadenine 9-beta-D-Arabinofuranoside 5'-Monophosphate((2R,3S,4S,5R)-5-(6-Amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

1763-50 1763-10
10188-076EA 93.39 USD
10188-076 10188-078
Fludarabine monophosphate
Fludarabine monophosphate
Formula: C₁₀H₁₃FN₅O₇P
MW: 365.21 g/mol
MDL Number: MFCD00866418
CAS Number: 75607-67-9

Order Now

Specification Test Results

CAS 75607-67-9
Purity ≥98%
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR