Triptolide
1761-5
1761-1
10188-008EA
89.98
USD
10188-008
10188-010
Triptolide
Triptolide
Formula:
C₂₀H₂₄O₆ |
MDL Number:
MFCD00210565 CAS Number: 38748-32-2 |
Specification Test Results
CAS | 38748-32-2 |
Purity | ≥97% by HPLC |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...