4-Nitroaniline ≥99%
Supplier: BioVision
Synonyms:
pNA
, p-Nitroaniline
AB145588-100MG
10191-618EA
183.6
USD
10191-618
4-Nitroaniline ≥99%
4-Nitroaniline
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00007858 CAS Number: 100-01-6 UN: 1661 ADR: 6.1,II |
Specification Test Results
CAS | 100-01-6 |
Purity |
|
Form | Golden yellow powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...