5-Aza-2'-deoxycytidine
Synonyms:
5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine
, Decitabine
, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one
1754-50
1754-10
10188-054EA
449.49
USD
10188-054
10188-052
5-Aza-2'-deoxycytidine
5-Aza-2'-deoxycytidine
Formula:
C₈H₁₂N₄O₄ MW: 228.21 g/mol |
MDL Number:
MFCD00043011 CAS Number: 2353-33-5 |
Specification Test Results
CAS | 2353-33-5 |
Purity | ≥98% by HPLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...